Assay Details
Gene Symbol: HOXB-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_102279.1 | 1 - 2 | 64 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | homeobox B2 | HOXB2 |
HOX2, HOX2H, Hox-2.8, K8 |
Anti-sense | Chr. 17 - 48542655 - 48545031 on NCBI Build GRCh38 | |
Downstream | homeobox B3 | HOXB3 |
HOX2, HOX2G, Hox-2.7 |
3.5kb | Anti-sense | Chr. 17 - 48548870 - 48590369 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000502764.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000502764.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000502764.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000502764.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000502764.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000502764.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000502764.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000504972.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000504972.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000504972.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | NR_102279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | NR_102279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | NR_102279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | NR_102279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | NR_102279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | NR_102279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | NR_102279.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700012278.hg.1 | NR_102279.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700202986.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700202986.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700202986.hg.1 | NR_102279.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700202986.hg.1 | TC1700012278.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700202986.hg.1 | NR_102279.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700202986.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700202986.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700202988.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700202988.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700202988.hg.1 | NR_102279.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700202988.hg.1 | TC1700012278.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700202988.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700202988.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700202988.hg.1 | NR_102279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000435312.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000502764.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000502764.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000502764.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000502764.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000502764.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000502764.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000502764.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000504972.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000504972.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000504972.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | ENST00000508688.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | NR_102279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | NR_102279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | NR_102279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | NR_102279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | NR_102279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | NR_102279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | NR_102279.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700012278.hg.1 | NR_102279.1 | Refseq |
