Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000502764.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000502764.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000502764.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000502764.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000502764.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000502764.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000502764.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000504972.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000504972.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000504972.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
NR_102279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
NR_102279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
NR_102279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
NR_102279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
NR_102279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
NR_102279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
NR_102279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012278.hg.1 |
NR_102279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700202990.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202990.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202990.hg.1 |
ENST00000504972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202990.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202990.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202990.hg.1 |
NR_102279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1700202990.hg.1 |
TC1700012278.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1700202990.hg.1 |
ENST00000504972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202990.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202990.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202990.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202990.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1700202990.hg.1 |
NR_102279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000435312.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000502764.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000502764.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000502764.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000502764.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000502764.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000502764.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000502764.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000504972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000504972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000504972.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
ENST00000508688.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
NR_102279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
NR_102279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
NR_102279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
NR_102279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
NR_102279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
NR_102279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
NR_102279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1700012278.hg.1 |
NR_102279.1 |
Refseq |