Assay Details
Gene Symbol: NRG1-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104156.1 | 2 - 3 | 256 | 114 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Werner syndrome RecQ like helicase | WRN |
RECQ3, RECQL2, RECQL3 |
-855.6kb | Sense | Chr. 8 - 31033262 - 31173761 on NCBI Build GRCh38 |
Downstream | NRG1 intronic transcript 3 | NRG1-IT3 | 410.3kb | Sense | Chr. 8 - 32440746 - 32442725 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000523743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000523743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000523743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000523743.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000523785.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000523785.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | ENST00000523785.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | NR_104156.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | NR_104156.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | NR_104156.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | NR_104156.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | NR_104156.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | NR_104156.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | NR_104156.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | NR_104157.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | NR_104157.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | NR_104157.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | NR_104157.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | NR_104157.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | NR_104157.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | NR_104158.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | NR_104158.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | NR_104158.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800007225.hg.1 | NR_104158.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000520444.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000521463.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000523743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000523743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000523743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000523743.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000523785.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000523785.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | ENST00000523785.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | NR_104156.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | NR_104156.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | NR_104156.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | NR_104156.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | NR_104156.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | NR_104156.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | NR_104156.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | NR_104157.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | NR_104157.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | NR_104157.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | NR_104157.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | NR_104157.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | NR_104157.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | NR_104158.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | NR_104158.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | NR_104158.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800007225.hg.1 | NR_104158.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800149719.hg.1 | ENST00000523743.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800149719.hg.1 | NR_104156.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800149719.hg.1 | TC0800007225.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800149719.hg.1 | NR_104156.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800149719.hg.1 | ENST00000523743.1 | Ensembl |
