Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000517411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000517411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000517411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000517411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000517411.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000518073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000518073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000518073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000518073.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000519852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000519852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000519852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000519852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000519852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000519852.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000521207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000521207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000521207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000521207.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000523031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000523031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000523031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000523031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000523031.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000524335.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000524335.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000524335.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000524335.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
ENST00000524335.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
NR_120682.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
NR_120682.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
NR_120682.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
NR_120682.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
NR_120682.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
NR_120683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
NR_120683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
NR_120683.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800012122.hg.1 |
NR_120683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000517411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000517411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000517411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000517411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000517411.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000518073.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000518073.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000518073.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000518073.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000519852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000519852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000519852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000519852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000519852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000519852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000521207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000521207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000521207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000521207.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000523031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000523031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000523031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000523031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000523031.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000524335.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000524335.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000524335.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000524335.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
ENST00000524335.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
NR_120682.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
NR_120682.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
NR_120682.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
NR_120682.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
NR_120682.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
NR_120683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
NR_120683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
NR_120683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800012122.hg.1 |
NR_120683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182485.hg.1 |
ENST00000518073.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182485.hg.1 |
ENST00000519852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182485.hg.1 |
ENST00000519852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182485.hg.1 |
NR_120683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182485.hg.1 |
NR_120683.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800182485.hg.1 |
TC0800012122.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800182485.hg.1 |
ENST00000518073.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182485.hg.1 |
ENST00000519852.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800182485.hg.1 |
ENST00000519852.1 |
Ensembl |