Assay Details
Gene Symbol: LOC101928279 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125892.1 | 3 | 1714 | 93 | ||
GenBank mRNA | AK056081.1 | - | 3 | 1670 | 93 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | STX18 antisense RNA 1 (head to head) | STX18-AS1 | -74.6kb | Sense | Chr. 4 - 4542131 - 4710938 on NCBI Build GRCh38 | |
Downstream | long intergenic non-protein coding RNA 1396 | LINC01396 | 57.1kb | Sense | Chr. 4 - 4844437 - 4850827 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
