Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
ENST00000521819.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
ENST00000521819.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
ENST00000521819.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
ENST00000521819.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
ENST00000521819.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
ENST00000523895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
ENST00000523895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
ENST00000523895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
ENST00000523895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
ENST00000523895.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200010254.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200172294.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172294.hg.1 |
ENST00000521819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172294.hg.1 |
ENST00000523895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172294.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200172294.hg.1 |
TC0200010254.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200172294.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200172294.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172294.hg.1 |
ENST00000521819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172294.hg.1 |
ENST00000523895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172296.hg.1 |
ENST00000523895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172296.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200172296.hg.1 |
TC0200010254.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200172296.hg.1 |
ENST00000523895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172296.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200172298.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172298.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200172298.hg.1 |
TC0200010254.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200172298.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172298.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
ENST00000521819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
ENST00000521819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
ENST00000521819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
ENST00000521819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
ENST00000521819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
ENST00000523895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
ENST00000523895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
ENST00000523895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
ENST00000523895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
ENST00000523895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200010254.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200172292.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172292.hg.1 |
ENST00000521819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172292.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200172292.hg.1 |
TC0200010254.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200172292.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172292.hg.1 |
ENST00000521819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172292.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200172293.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172293.hg.1 |
ENST00000521819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172293.hg.1 |
ENST00000523895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172293.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200172293.hg.1 |
TC0200010254.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200172293.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172293.hg.1 |
ENST00000521819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172293.hg.1 |
ENST00000523895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172293.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200172295.hg.1 |
ENST00000521819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172295.hg.1 |
ENST00000523895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172295.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200172295.hg.1 |
TC0200010254.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200172295.hg.1 |
ENST00000521819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172295.hg.1 |
ENST00000523895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172295.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200172297.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200172297.hg.1 |
TC0200010254.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200172297.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200172291.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172291.hg.1 |
ENST00000521819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172291.hg.1 |
ENST00000523895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172291.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200172291.hg.1 |
TC0200010254.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200172291.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172291.hg.1 |
ENST00000521819.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172291.hg.1 |
ENST00000523895.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172291.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200172289.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172289.hg.1 |
NR_102429.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200172289.hg.1 |
TC0200010254.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200172289.hg.1 |
ENST00000520651.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200172289.hg.1 |
NR_102429.1 |
Refseq |