Assay Details
Gene Symbol: AR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FJ235916.1 | - | 4 | 2260 | 62 | |
FJ235919.1 | - | 4 | 2658 | 62 | ||
FJ235920.1 | - | 5 | 2541 | 62 | ||
HM055487.1 | - | 3 | 2177 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
