Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017289.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017289.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017289.hg.1 |
TC10001366.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000010925.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017291.hg.1 |
ENST00000318971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017291.hg.1 |
ENST00000373209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017291.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017291.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017291.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017291.hg.1 |
TC10001366.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017290.hg.1 |
ENST00000318971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017290.hg.1 |
ENST00000373209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017290.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017290.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017290.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017290.hg.1 |
TC10001366.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017287.hg.1 |
ENST00000318971.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017287.hg.1 |
ENST00000373209.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017287.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017287.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017287.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017287.hg.1 |
TC10001366.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000174383.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174383.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174383.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174383.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174383.hg.1 |
TC1000010925.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000174383.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174383.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174383.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174383.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174380.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174380.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174380.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174380.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174380.hg.1 |
TC1000010925.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000174380.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174380.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174380.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174380.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174382.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174382.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174382.hg.1 |
TC1000010925.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000174382.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174382.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000010925.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174384.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174384.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174384.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174384.hg.1 |
NM_005041.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174384.hg.1 |
TC1000010925.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000174384.hg.1 |
ENST00000373209.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174384.hg.1 |
ENST00000441259.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000174384.hg.1 |
NM_001083116.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000174384.hg.1 |
NM_005041.5 |
Refseq |