Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
ENST00000260442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
ENST00000260442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
ENST00000260442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
ENST00000260442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
ENST00000260442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
ENST00000260442.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
ENST00000561198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
ENST00000561198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
ENST00000561198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
ENST00000561198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
ENST00000561198.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
NM_001306168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
NM_001306168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
NM_001306168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
NM_001306168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
NM_001306168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
NM_001306168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
NM_001306168.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
NM_020396.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
NM_020396.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
NM_020396.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
NM_020396.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
NM_020396.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009451.hg.1 |
NM_020396.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
ENST00000260442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
ENST00000260442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
ENST00000260442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
ENST00000260442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
ENST00000260442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
ENST00000260442.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
ENST00000561198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
ENST00000561198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
ENST00000561198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
ENST00000561198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
ENST00000561198.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
NM_001306168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
NM_001306168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
NM_001306168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
NM_001306168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
NM_001306168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
NM_001306168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
NM_001306168.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
NM_020396.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
NM_020396.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
NM_020396.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
NM_020396.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
NM_020396.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500009451.hg.1 |
NM_020396.3 |
Refseq |