Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000435649.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000435649.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000435649.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000435649.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000443802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000443802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000443802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000443802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000443802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000443802.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000606660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000606660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000606660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000606660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000606660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000606660.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100014891.hg.1 |
ENST00000606660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000435649.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000435649.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000435649.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000435649.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000443802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000443802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000443802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000443802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000443802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000443802.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000455680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000606660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000606660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000606660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000606660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000606660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000606660.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100014891.hg.1 |
ENST00000606660.1 |
Ensembl |