Assay Details
Gene Symbol: LOC102723828 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_125936.1 | 1 - 2 | 439 | 103 | ||
GenBank mRNA | CX756508.1 | - | 1 - 2 | 422 | 103 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC105377651 | LOC105377651 | Anti-sense | Chr. 4 - 32005853 - 32022422 on NCBI Build GRCh38 | ||
Downstream | uncharacterized LOC101927363 | LOC101927363 | 204.8kb | Sense | Chr. 4 - 32351038 - 32353220 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
