Assay Details
Gene Symbol: LINC01044 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_126345.1 | 1 - 2 | 163 | 60 | ||
GenBank mRNA | BG722997.1 | - | 1 - 2 | 167 | 60 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | long intergenic non-protein coding RNA 1043 | LINC01043 | -5.5kb | Anti-sense | Chr. 13 - 112313467 - 112321766 on NCBI Build GRCh38 | |
Downstream | sperm acrosome associated 7 | SPACA7 | C13orf28 | 45.1kb | Anti-sense | Chr. 13 - 112376300 - 112434695 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
