Assay Details
Gene Symbol: NCOA7-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_126386.1 | 1 - 2 | 47 | 98 | ||
GenBank mRNA | AW444508.1 | - | 2 - 3 | 98 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nuclear receptor coactivator 7 | NCOA7 |
ERAP140, ESNA1, NCOA7-AS, Nbla00052, Nbla10993, TLDC4, dJ187J11.3 |
Anti-sense | Chr. 6 - 125780948 - 125932030 on NCBI Build GRCh38 | |
Downstream | histidine triad nucleotide binding protein 3 | HINT3 | HINT4 | 137.9kb | Anti-sense | Chr. 6 - 125956715 - 125980244 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
