Assay Details
Gene Symbol: LINC01588 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_126500.1 | 2 - 3 | 1507 | 58 | ||
GenBank mRNA | CR980193.1 | - | 2 - 3 | 228 | 58 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 6076 | MIR6076 | hsa-mir-6076 | -32.6kb | Anti-sense | Chr. 14 - 49966399 - 49966511 on NCBI Build GRCh38 |
Downstream | long intergenic non-protein coding RNA 1599 | LINC01599 | C14orf183 | 4.4kb | Sense | Chr. 14 - 50007313 - 50105043 on NCBI Build GRCh38 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000399206.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000399206.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000399206.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000399206.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000399206.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000399206.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000399206.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000399206.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000399206.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000399206.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000530176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000530176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000530176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000530176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000530176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000530176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000530176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000530176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000530176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000530176.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000533506.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000533506.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000533506.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000533506.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000533506.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000533506.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | ENST00000533506.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126497.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126497.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126497.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126497.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126497.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126497.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126497.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126497.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126499.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126500.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126500.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126500.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126500.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126500.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126500.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126500.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126500.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126500.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010748.hg.1 | NR_126500.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000399206.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000399206.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000399206.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000399206.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000399206.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000399206.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000399206.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000399206.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000399206.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000399206.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000528300.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000529902.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000530176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000530176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000530176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000530176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000530176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000530176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000530176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000530176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000530176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000530176.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000533506.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000533506.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000533506.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000533506.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000533506.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000533506.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | ENST00000533506.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126497.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126497.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126497.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126497.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126497.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126497.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126497.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126497.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126498.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126499.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126500.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126500.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126500.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126500.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126500.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126500.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126500.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126500.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126500.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010748.hg.1 | NR_126500.1 | Refseq |
