Assay Details
Gene Symbol: C16orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011522462.2 | XP_011520764.1 | 4 | 1788 | 106 | |
GenBank mRNA | AK095835.1 | - | 1 | 516 | 106 | |
AL832650.1 | - | 1 | 1619 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000327827.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574285.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | ENST00000574616.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600006842.hg.1 | NM_014117.2 | Refseq |
