Assay Details
Gene Symbol: LOC101927865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DB075298.1 | - | 1 - 2 | 84 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
