Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015912.hg.1 |
ENST00000305123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015912.hg.1 |
ENST00000305123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015912.hg.1 |
ENST00000305123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015912.hg.1 |
ENST00000305123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015912.hg.1 |
ENST00000305123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015912.hg.1 |
ENST00000305123.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015912.hg.1 |
ENST00000498335.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015912.hg.1 |
ENST00000498335.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015912.hg.1 |
NM_005544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015912.hg.1 |
NM_005544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015912.hg.1 |
NM_005544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015912.hg.1 |
NM_005544.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200015912.hg.1 |
NM_005544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015912.hg.1 |
ENST00000305123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015912.hg.1 |
ENST00000305123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015912.hg.1 |
ENST00000305123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015912.hg.1 |
ENST00000305123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015912.hg.1 |
ENST00000305123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015912.hg.1 |
ENST00000305123.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015912.hg.1 |
ENST00000498335.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015912.hg.1 |
ENST00000498335.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200015912.hg.1 |
NM_005544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015912.hg.1 |
NM_005544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015912.hg.1 |
NM_005544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015912.hg.1 |
NM_005544.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200015912.hg.1 |
NM_005544.2 |
Refseq |