Assay Details
Gene Symbol: CREB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004379.4 | NP_004370.1 | 8 | 7615 | 124 | |
NM_134442.4 | NP_604391.1 | 9 | 7657 | 124 | ||
NR_135473.1 | 10 | 6061 | 124 | |||
GenBank mRNA | AK091534.1 | - | 1 | 821 | 124 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
