Assay Details
Gene Symbol: LOC644838 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038844.1 | 1 - 2 | 91 | 100 | ||
GenBank mRNA | AK123766.1 | - | 1 - 2 | 91 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000414404.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000414404.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000414404.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000414404.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000414404.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000447453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000447453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000447453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | ENST00000447453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | NR_038844.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | NR_038844.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | NR_038844.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | NR_038844.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | NR_038844.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | NR_038844.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | NR_038844.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | NR_038844.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | NR_038844.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200012908.hg.1 | NR_038844.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000414404.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000414404.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000414404.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000414404.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000414404.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000433133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000447453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000447453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000447453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | ENST00000447453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | NR_038844.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | NR_038844.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | NR_038844.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | NR_038844.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | NR_038844.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | NR_038844.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | NR_038844.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | NR_038844.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | NR_038844.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200012908.hg.1 | NR_038844.1 | Refseq |
