Assay Details
Gene Symbol: CCDC13-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_046732.1 | 3 - 4 | 336 | 115 | ||
GenBank mRNA | AK098828.1 | - | 2 - 3 | 235 | 115 | |
DB179860.1 | - | 3 - 4 | 339 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000435327.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000435327.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000435327.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000435327.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000435327.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000435327.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000435327.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000466031.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000466031.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000466031.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000472921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000472921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000472921.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000479576.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000479576.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000479576.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000479576.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000479576.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000479576.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000492806.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000492806.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000492806.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000492806.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000492806.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000492806.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000492806.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | ENST00000492806.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007170.hg.1 | ENST00000418161.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007170.hg.1 | ENST00000418161.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007170.hg.1 | ENST00000418161.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007170.hg.1 | ENST00000418161.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007170.hg.1 | ENST00000446950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007170.hg.1 | ENST00000446950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007170.hg.1 | ENST00000446950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007170.hg.1 | ENST00000446950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007170.hg.1 | ENST00000446950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007170.hg.1 | ENST00000446950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007170.hg.1 | ENST00000446950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007170.hg.1 | ENST00000446950.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300007170.hg.1 | NR_046732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007170.hg.1 | NR_046732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007170.hg.1 | NR_046732.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300007170.hg.1 | NR_046732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007170.hg.1 | ENST00000418161.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007170.hg.1 | ENST00000418161.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007170.hg.1 | ENST00000418161.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007170.hg.1 | ENST00000418161.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007170.hg.1 | ENST00000446950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007170.hg.1 | ENST00000446950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007170.hg.1 | ENST00000446950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007170.hg.1 | ENST00000446950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007170.hg.1 | ENST00000446950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007170.hg.1 | ENST00000446950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007170.hg.1 | ENST00000446950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007170.hg.1 | ENST00000446950.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300007170.hg.1 | NR_046732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007170.hg.1 | NR_046732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007170.hg.1 | NR_046732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300007170.hg.1 | NR_046732.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000310232.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000435327.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000435327.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000435327.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000435327.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000435327.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000435327.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000435327.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000466031.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000466031.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000466031.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000472921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000472921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000472921.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000479576.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000479576.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000479576.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000479576.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000479576.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000479576.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000492806.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000492806.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000492806.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000492806.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000492806.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000492806.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000492806.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | ENST00000492806.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | NM_144719.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013959.hg.1 | NM_144719.3 | Refseq |
