Assay Details
Gene Symbol: ZBTB11-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_024407.1 | 1 | 1116 | 70 | ||
GenBank mRNA | AK022880.1 | - | 1 | 1100 | 70 | |
BC003066.2 | - | 1 | 1116 | 70 | ||
BC016930.1 | - | 1 | 1071 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000364251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000461821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000461821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000461821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000461821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000461821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000461821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000461821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000471673.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000471673.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | ENST00000471673.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008176.hg.1 | ENST00000536865.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008176.hg.1 | ENST00000536865.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008176.hg.1 | ENST00000609682.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008176.hg.1 | ENST00000609682.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008176.hg.1 | NR_024407.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008176.hg.1 | NR_024407.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008176.hg.1 | NR_024407.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008176.hg.1 | ENST00000536865.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008176.hg.1 | ENST00000536865.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008176.hg.1 | ENST00000609682.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008176.hg.1 | ENST00000609682.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008176.hg.1 | NR_024407.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008176.hg.1 | NR_024407.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008176.hg.1 | NR_024407.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000312938.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000364251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000461821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000461821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000461821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000461821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000461821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000461821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000461821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000471673.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000471673.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | ENST00000471673.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | NM_014415.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300011859.hg.1 | NM_014415.3 | Refseq |
