Assay Details
Gene Symbol: HEG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005247666.1 | XP_005247723.1 | 6 - 7 | 1950 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
vasculogenesis
in utero embryonic development
endothelial cell morphogenesis
lymph vessel development
lymph circulation
cardiac atrium morphogenesis
ventricular trabecula myocardium morphogenesis
ventricular septum development
cell-cell junction assembly
post-embryonic development
lung development
multicellular organism growth
venous blood vessel morphogenesis
regulation of body fluid levels
cardiac muscle tissue growth
pericardium development
positive regulation of fibroblast growth factor production
calcium ion binding
vasculogenesis
in utero embryonic development
endothelial cell morphogenesis
lymph vessel development
lymph circulation
cardiac atrium morphogenesis
ventricular trabecula myocardium morphogenesis
ventricular septum development
cell-cell junction assembly
post-embryonic development
lung development
multicellular organism growth
venous blood vessel morphogenesis
regulation of body fluid levels
cardiac muscle tissue growth
pericardium development
positive regulation of fibroblast growth factor production
calcium ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000477536.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000477536.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000477536.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000480667.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000480667.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000480667.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000480667.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000482699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000482699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000482699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000482699.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000487661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000487661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000487661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000487661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000487661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000487661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000487661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000487661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000487661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000487661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000488654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000488654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | ENST00000488654.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000311127.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000477536.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000477536.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000477536.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000480667.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000480667.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000480667.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000480667.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000482699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000482699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000482699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000482699.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000487661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000487661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000487661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000487661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000487661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000487661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000487661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000487661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000487661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000487661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000488654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000488654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | ENST00000488654.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300012238.hg.1 | NM_020733.1 | Refseq |
