Assay Details
Gene Symbol: SDHAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK297840.1 | - | 1 - 2 | 301 | 58 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK297840.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000385056.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000385194.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000399540.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000399540.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000399540.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000399540.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000399540.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000399541.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000399541.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000399541.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000399541.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000443997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000443997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000443997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000443997.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000570562.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000570562.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000570562.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000570562.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000570562.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000570562.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000571010.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000571010.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000571010.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000571010.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572385.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572385.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572385.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572385.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572385.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572385.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000572547.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000573939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000573939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000573939.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000574377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000574377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000574377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000574377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000574377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000574377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000574377.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000575727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000575727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000575727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000575727.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000575889.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000575889.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000575889.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000575889.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | ENST00000575889.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | NR_029712.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | NR_030178.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | NR_038310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | NR_038310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | NR_038310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | NR_038310.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | NR_040089.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | NR_040089.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | NR_040089.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700009602.hg.1 | NR_040089.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013284.hg.1 | ENST00000436493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013284.hg.1 | ENST00000436493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013284.hg.1 | ENST00000436493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013284.hg.1 | ENST00000436493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013284.hg.1 | ENST00000436493.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013284.hg.1 | ENST00000515467.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013284.hg.1 | ENST00000515467.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013284.hg.1 | ENST00000515467.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013284.hg.1 | ENST00000515467.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013284.hg.1 | ENST00000515467.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013284.hg.1 | ENST00000515467.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013284.hg.1 | ENST00000515467.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500013284.hg.1 | NR_003263.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013284.hg.1 | NR_003263.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013284.hg.1 | NR_003263.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013284.hg.1 | NR_003263.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013284.hg.1 | NR_003263.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013284.hg.1 | NR_003263.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013284.hg.1 | NR_003263.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013284.hg.1 | NR_003263.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500013284.hg.1 | NR_003263.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000385056.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000385194.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000399540.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000399540.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000399540.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000399540.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000399540.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000399541.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000399541.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000399541.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000399541.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000443997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000443997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000443997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000443997.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000570562.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000570562.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000570562.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000570562.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000570562.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000570562.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000571010.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000571010.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000571010.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000571010.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572385.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572385.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572385.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572385.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572385.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572385.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572547.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572547.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572547.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000572547.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000573939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000573939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000573939.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000574377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000574377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000574377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000574377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000574377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000574377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000574377.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000575727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000575727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000575727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000575727.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000575889.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000575889.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000575889.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000575889.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | ENST00000575889.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | NR_029712.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | NR_030178.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | NR_038310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | NR_038310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | NR_038310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | NR_038310.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | NR_040089.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | NR_040089.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | NR_040089.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700009602.hg.1 | NR_040089.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013284.hg.1 | ENST00000436493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013284.hg.1 | ENST00000436493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013284.hg.1 | ENST00000436493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013284.hg.1 | ENST00000436493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013284.hg.1 | ENST00000436493.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013284.hg.1 | ENST00000515467.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013284.hg.1 | ENST00000515467.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013284.hg.1 | ENST00000515467.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013284.hg.1 | ENST00000515467.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013284.hg.1 | ENST00000515467.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013284.hg.1 | ENST00000515467.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013284.hg.1 | ENST00000515467.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500013284.hg.1 | NR_003263.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013284.hg.1 | NR_003263.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013284.hg.1 | NR_003263.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013284.hg.1 | NR_003263.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013284.hg.1 | NR_003263.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013284.hg.1 | NR_003263.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013284.hg.1 | NR_003263.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013284.hg.1 | NR_003263.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0500013284.hg.1 | NR_003263.2 | Refseq |
