Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012961.hg.1 |
ENST00000295663.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012961.hg.1 |
ENST00000295663.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012961.hg.1 |
ENST00000295663.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012961.hg.1 |
ENST00000527261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012961.hg.1 |
ENST00000527261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012961.hg.1 |
ENST00000527261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012961.hg.1 |
ENST00000527261.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012961.hg.1 |
ENST00000527369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012961.hg.1 |
ENST00000527369.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012961.hg.1 |
ENST00000532966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012961.hg.1 |
ENST00000532966.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012961.hg.1 |
NR_103765.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012961.hg.1 |
NR_103765.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012961.hg.1 |
NR_103765.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012961.hg.1 |
NR_103765.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012961.hg.1 |
NR_103765.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012961.hg.1 |
NR_103765.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100012961.hg.1 |
NR_103765.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012961.hg.1 |
ENST00000295663.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012961.hg.1 |
ENST00000295663.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012961.hg.1 |
ENST00000295663.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012961.hg.1 |
ENST00000527261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012961.hg.1 |
ENST00000527261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012961.hg.1 |
ENST00000527261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012961.hg.1 |
ENST00000527261.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012961.hg.1 |
ENST00000527369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012961.hg.1 |
ENST00000527369.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012961.hg.1 |
ENST00000532966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012961.hg.1 |
ENST00000532966.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100012961.hg.1 |
NR_103765.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012961.hg.1 |
NR_103765.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012961.hg.1 |
NR_103765.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012961.hg.1 |
NR_103765.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012961.hg.1 |
NR_103765.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012961.hg.1 |
NR_103765.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100012961.hg.1 |
NR_103765.1 |
Refseq |