Assay Details
Gene Symbol: BBOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003986.2 | NP_003977.1 | 9 | 1654 | 138 | |
XM_005253159.2 | XP_005253216.1 | 9 | 1902 | 138 | ||
XM_005253160.2 | XP_005253217.1 | 9 | 1899 | 138 | ||
XM_005253161.2 | XP_005253218.1 | 9 | 1651 | 138 | ||
XM_011520402.1 | XP_011518704.1 | 10 | 2062 | 138 | ||
XM_017018403.1 | XP_016873892.1 | 8 | 1584 | 138 | ||
GenBank mRNA | AF082868.1 | - | 8 | 1352 | 138 | |
AK290108.1 | - | 8 | 1399 | 138 | ||
BC011034.1 | - | 7 | 1342 | 4042355 | 138 | |
DQ471869.1 | - | 1 | 125 | 138 | ||
DQ471870.1 | - | 1 | 125 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000534323.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000534323.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000534323.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | ENST00000534323.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000263182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000525090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000527505.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000528583.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000529202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000531832.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000533566.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000534323.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000534323.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000534323.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | ENST00000534323.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007111.hg.1 | NM_003986.2 | Refseq |
