Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
ENST00000455530.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014146.hg.1 |
NR_125859.1 |
Refseq |