Assay Details
Gene Symbol: LOC101928738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BM850955.1 | - | 1 - 2 | 206 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
