Assay Details
Gene Symbol: UST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011535378.2 | XP_011533680.1 | 5 - 6 | 1668 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | ENST00000466695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | ENST00000466695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | ENST00000466695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | ENST00000473631.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | ENST00000473631.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | ENST00000473631.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | ENST00000367463.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | ENST00000466695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | ENST00000466695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | ENST00000466695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | ENST00000473631.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | ENST00000473631.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | ENST00000473631.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | NM_005715.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600009783.hg.1 | NM_005715.2 | Refseq |
