Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000300303.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000300303.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000300303.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000300303.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000300303.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000300303.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000300303.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000300303.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000443096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000443096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000443096.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000444446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000444446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000444446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000444446.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000455377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000455377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000455377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000455377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000455377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000455377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
ENST00000455377.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
NM_198571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
NM_198571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
NM_198571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
NM_198571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
NM_198571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
NM_198571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
NM_198571.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012037.hg.1 |
NM_198571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000300303.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000300303.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000300303.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000300303.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000300303.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000300303.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000300303.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000300303.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000443096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000443096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000443096.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000444446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000444446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000444446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000444446.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000455377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000455377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000455377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000455377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000455377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000455377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
ENST00000455377.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
NM_198571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
NM_198571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
NM_198571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
NM_198571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
NM_198571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
NM_198571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
NM_198571.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012037.hg.1 |
NM_198571.2 |
Refseq |