Assay Details
Gene Symbol: MINCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AA284855.1 | - | 1 | 92 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000517411.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000517411.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000517411.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000517411.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000517411.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000518073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000518073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000518073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000518073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000519852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000519852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000519852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000519852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000519852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000519852.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000521207.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000521207.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000521207.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000521207.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000523031.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000523031.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000523031.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000523031.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000523031.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000524335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000524335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000524335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000524335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | ENST00000524335.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | NR_120682.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | NR_120682.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | NR_120682.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | NR_120682.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | NR_120682.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | NR_120683.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | NR_120683.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | NR_120683.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800012122.hg.1 | NR_120683.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000517411.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000517411.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000517411.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000517411.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000517411.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000518073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000518073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000518073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000518073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000519852.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000519852.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000519852.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000519852.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000519852.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000519852.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000521207.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000521207.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000521207.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000521207.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000523031.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000523031.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000523031.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000523031.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000523031.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000524335.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000524335.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000524335.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000524335.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | ENST00000524335.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | NR_120682.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | NR_120682.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | NR_120682.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | NR_120682.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | NR_120682.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | NR_120683.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | NR_120683.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | NR_120683.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800012122.hg.1 | NR_120683.1 | Refseq |
