Assay Details
Gene Symbol: BAALC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK310816.1 | - | 1 - 2 | 267 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000306391.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000306391.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000330955.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000330955.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000517863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000517863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000517863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000519507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000519507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000519507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000519507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000521926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000521926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000521926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000521926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000521926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000297574.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000306391.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000306391.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000309982.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000330955.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000330955.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000438105.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000517863.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000517863.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000517863.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000519507.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000519507.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000519507.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000519507.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000521926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000521926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000521926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000521926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000521926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | ENST00000523754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_001024372.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0800008474.hg.1 | NM_024812.2 | Refseq |
