Assay Details
Gene Symbol: DCAF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286810.1 | NP_001273739.1 | 7 | 6379 | 96 | |
NM_024345.4 | NP_077321.3 | 7 | 6905 | 96 | ||
XM_017015129.1 | XP_016870618.1 | 6 | 6215 | 96 | ||
GenBank mRNA | AK023620.1 | - | 1 | 1627 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000452964.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000242323.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000377724.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000452964.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000461549.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000478453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | ENST00000483167.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_001286810.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900007167.hg.1 | NM_024345.4 | Refseq |
