Assay Details
Gene Symbol: CHRNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006720382.2 | XP_006720445.1 | 1 - 2 | 522 | 56 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006720382.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
ion transport
regulation of smooth muscle contraction
signal transduction
activation of transmembrane receptor protein tyrosine kinase activity
synaptic transmission, cholinergic
nervous system development
heart development
locomotory behavior
response to xenobiotic stimulus
response to inorganic substance
regulation of acetylcholine secretion, neurotransmission
behavioral response to nicotine
regulation of membrane potential
regulation of dendrite morphogenesis
protein heterooligomerization
excitatory postsynaptic potential
synaptic transmission involved in micturition
cation transmembrane transport
ligand-gated ion channel activity
acetylcholine receptor activity
acetylcholine-gated cation-selective channel activity
acetylcholine binding
protein heterodimerization activity
ion transport
regulation of smooth muscle contraction
signal transduction
activation of transmembrane receptor protein tyrosine kinase activity
synaptic transmission, cholinergic
nervous system development
heart development
locomotory behavior
response to xenobiotic stimulus
response to inorganic substance
regulation of acetylcholine secretion, neurotransmission
behavioral response to nicotine
regulation of membrane potential
regulation of dendrite morphogenesis
protein heterooligomerization
excitatory postsynaptic potential
synaptic transmission involved in micturition
cation transmembrane transport
ligand-gated ion channel activity
acetylcholine receptor activity
acetylcholine-gated cation-selective channel activity
acetylcholine binding
protein heterodimerization activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000558903.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000558903.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559002.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559002.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559002.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559002.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559080.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559080.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559080.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559080.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559941.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559941.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000561128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000561128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000561128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000561128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000561128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000558903.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000558903.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559002.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559002.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559002.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559002.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559080.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559080.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559080.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559080.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559941.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559941.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000561128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000561128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000561128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000561128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000561128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
