Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009346.hg.1 |
ENST00000467564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009346.hg.1 |
ENST00000467564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009346.hg.1 |
ENST00000467564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009346.hg.1 |
ENST00000467564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009346.hg.1 |
ENST00000467564.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009346.hg.1 |
ENST00000497071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009346.hg.1 |
ENST00000497071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009346.hg.1 |
ENST00000497071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009346.hg.1 |
ENST00000497071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009346.hg.1 |
ENST00000497071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009346.hg.1 |
ENST00000497071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009346.hg.1 |
ENST00000497071.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200009346.hg.1 |
ENST00000497071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009346.hg.1 |
ENST00000467564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009346.hg.1 |
ENST00000467564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009346.hg.1 |
ENST00000467564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009346.hg.1 |
ENST00000467564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009346.hg.1 |
ENST00000467564.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009346.hg.1 |
ENST00000497071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009346.hg.1 |
ENST00000497071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009346.hg.1 |
ENST00000497071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009346.hg.1 |
ENST00000497071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009346.hg.1 |
ENST00000497071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009346.hg.1 |
ENST00000497071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009346.hg.1 |
ENST00000497071.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200009346.hg.1 |
ENST00000497071.1 |
Ensembl |