Assay Details
Gene Symbol: ADORA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK308733.1 | - | 2 - 3 | 450 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000464019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000464019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000464019.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000467253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000467253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000467253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000467253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000467253.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000472535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000472535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000472535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000472535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000472535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000472535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000618295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000618295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000618295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000618295.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367235.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000464019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000464019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000464019.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000467253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000467253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000467253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000467253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000467253.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000472535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000472535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000472535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000472535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000472535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000472535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000618295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000618295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000618295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000618295.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_000674.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | NM_001048230.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000309502.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000337894.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367235.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100011245.hg.1 | ENST00000367236.8 | Ensembl |
