Assay Details
Gene Symbol: CYP24A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005260304.4 | XP_005260361.1 | 11 - 12 | 1963 | 131 | |
XM_017027693.1 | XP_016883182.1 | 10 - 11 | 1765 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
osteoblast differentiation
vitamin metabolic process
response to vitamin D
vitamin D metabolic process
vitamin D catabolic process
vitamin D receptor signaling pathway
iron ion binding
25-hydroxycholecalciferol-24-hydroxylase activity
oxidoreductase activity
heme binding
1-alpha,25-dihydroxyvitamin D3 24-hydroxylase activity
osteoblast differentiation
vitamin metabolic process
response to vitamin D
vitamin D metabolic process
vitamin D catabolic process
vitamin D receptor signaling pathway
iron ion binding
25-hydroxycholecalciferol-24-hydroxylase activity
oxidoreductase activity
heme binding
1-alpha,25-dihydroxyvitamin D3 24-hydroxylase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000460643.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000460643.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000460643.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000460643.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000472970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000472970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000472970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000487593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000487593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | ENST00000487593.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000216862.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395954.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000395955.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000460643.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000460643.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000460643.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000460643.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000472970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000472970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000472970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000487593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000487593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | ENST00000487593.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_000782.4 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2000009522.hg.1 | NM_001128915.1 | Refseq |
