Assay Details
Gene Symbol: EPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015630.3 | NP_056445.3 | 1 - 2 | 178 | 107 | |
XM_011510941.2 | XP_011509243.1 | 1 - 2 | 389 | 107 | ||
GenBank mRNA | AF286904.1 | - | 1 - 2 | 170 | 107 | |
AK125789.1 | - | 2 - 3 | 247 | 107 | ||
BC093818.1 | - | 1 - 2 | 90 | 7939663 | 107 | |
BC093820.1 | - | 1 - 2 | 90 | 7939665 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF286904.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000449013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000449013.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000258484.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000397424.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000409654.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000449013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000449013.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000457184.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | ENST00000491099.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200009612.hg.1 | NM_015630.3 | Refseq |
