Assay Details
Gene Symbol: CASC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026939.1 | 5 | 804 | 77 | ||
NR_026940.1 | 4 | 704 | 77 | |||
GenBank mRNA | AJ535621.1 | - | 4 | 517 | 77 | |
AY226450.1 | - | 5 | 620 | 77 | ||
BC036393.1 | - | 5 | 805 | 77 | ||
BC112306.1 | - | 4 | 528 | 77 | ||
BC112308.1 | - | 4 | 528 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000426021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000426021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000426021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000426021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000426021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000426021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000426021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000426021.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000439517.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000439517.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000439517.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000439517.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454781.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000614455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000614455.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000622752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | ENST00000622752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026939.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026940.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026940.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026940.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026940.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026940.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026940.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026940.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026940.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026941.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026941.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026941.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026941.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026941.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009037.hg.1 | NR_026941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000414722.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000426021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000426021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000426021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000426021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000426021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000426021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000426021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000426021.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000435944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000439517.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000439517.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000439517.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000439517.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454781.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000454857.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000614455.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000614455.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000622752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | ENST00000622752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026939.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026940.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026940.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026940.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026940.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026940.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026940.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026940.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026940.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026941.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009037.hg.1 | NR_026941.1 | Refseq |
