Assay Details
Gene Symbol: PKD1P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK310228.1 | - | 8 | 2124 | 89 |
Gene Symbol: PKD1P6-NPIPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK303260.1 | - | 8 | 1630 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
