Assay Details
Gene Symbol: LOC105376037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK098263.1 | - | 2 | 317 | 102 | |
AK126527.1 | - | 1 | 1666 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400007058.hg.1 | ENST00000503017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007058.hg.1 | ENST00000503017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007058.hg.1 | ENST00000503017.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007058.hg.1 | ENST00000503394.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007058.hg.1 | ENST00000503394.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007058.hg.1 | ENST00000503394.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007058.hg.1 | ENST00000503394.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007058.hg.1 | ENST00000503394.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007058.hg.1 | ENST00000503394.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007058.hg.1 | ENST00000506425.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007058.hg.1 | ENST00000506425.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007058.hg.1 | ENST00000506425.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007058.hg.1 | ENST00000506425.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007058.hg.1 | ENST00000511453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007058.hg.1 | ENST00000511453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007058.hg.1 | ENST00000511453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007058.hg.1 | ENST00000511453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007058.hg.1 | ENST00000511453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007058.hg.1 | ENST00000511453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007058.hg.1 | ENST00000511453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007058.hg.1 | ENST00000511453.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000327304.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000327304.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000327304.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000327304.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000327304.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000327304.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000327304.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000327304.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000327304.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000327304.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000396521.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000396521.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000396521.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000396521.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000396521.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000396521.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000465229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000465229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000465229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000465229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000465229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000465229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000465229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000465229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000465229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000465860.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000465860.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000465860.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000465860.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000482614.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000482614.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000482614.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000482614.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000489414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000489414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000489414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000489414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000489414.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000490516.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000490516.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000490516.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000490516.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000490516.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000496910.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000496910.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | ENST00000496910.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | NM_001002269.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | NM_001002269.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | NM_001002269.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | NM_001002269.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | NM_001002269.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | NM_001002269.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | NM_001002269.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | NM_001002269.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | NM_001002269.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | NM_001002269.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | NM_016042.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | NM_016042.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | NM_016042.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | NM_016042.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | NM_016042.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | NM_016042.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | NM_016042.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | NM_016042.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | NM_016042.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | NM_016042.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900012229.hg.1 | NM_016042.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007058.hg.1 | ENST00000503017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007058.hg.1 | ENST00000503017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007058.hg.1 | ENST00000503017.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007058.hg.1 | ENST00000503394.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007058.hg.1 | ENST00000503394.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007058.hg.1 | ENST00000503394.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007058.hg.1 | ENST00000503394.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007058.hg.1 | ENST00000503394.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007058.hg.1 | ENST00000503394.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007058.hg.1 | ENST00000506425.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007058.hg.1 | ENST00000506425.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007058.hg.1 | ENST00000506425.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007058.hg.1 | ENST00000506425.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007058.hg.1 | ENST00000511453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007058.hg.1 | ENST00000511453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007058.hg.1 | ENST00000511453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007058.hg.1 | ENST00000511453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007058.hg.1 | ENST00000511453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007058.hg.1 | ENST00000511453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007058.hg.1 | ENST00000511453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007058.hg.1 | ENST00000511453.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000327304.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000327304.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000327304.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000327304.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000327304.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000327304.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000327304.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000327304.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000327304.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000327304.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000396521.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000396521.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000396521.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000396521.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000396521.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000396521.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000465229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000465229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000465229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000465229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000465229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000465229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000465229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000465229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000465229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000465860.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000465860.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000465860.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000465860.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000482614.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000482614.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000482614.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000482614.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000489414.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000489414.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000489414.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000489414.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000489414.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000490516.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000490516.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000490516.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000490516.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000490516.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000496910.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000496910.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | ENST00000496910.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | NM_001002269.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | NM_001002269.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | NM_001002269.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | NM_001002269.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | NM_001002269.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | NM_001002269.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | NM_001002269.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | NM_001002269.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | NM_001002269.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | NM_001002269.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | NM_016042.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | NM_016042.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | NM_016042.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | NM_016042.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | NM_016042.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | NM_016042.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | NM_016042.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | NM_016042.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | NM_016042.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | NM_016042.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0900012229.hg.1 | NM_016042.3 | Refseq |
