Assay Details
Gene Symbol: KRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017019292.1 | XP_016874781.1 | 3 | 1284 | 103 | |
GenBank mRNA | BC029545.1 | - | 3 | 723 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
positive regulation of protein phosphorylation
stimulatory C-type lectin receptor signaling pathway
epidermal growth factor receptor signaling pathway
Ras protein signal transduction
axon guidance
positive regulation of cell population proliferation
visual learning
positive regulation of gene expression
cytokine-mediated signaling pathway
forebrain astrocyte development
actin cytoskeleton organization
regulation of protein stability
regulation of synaptic transmission, GABAergic
positive regulation of Rac protein signal transduction
social behavior
endocrine signaling
Fc-epsilon receptor signaling pathway
ERBB2 signaling pathway
positive regulation of MAP kinase activity
negative regulation of neuron apoptotic process
negative regulation of cell differentiation
regulation of long-term neuronal synaptic plasticity
homeostasis of number of cells within a tissue
leukocyte migration
positive regulation of nitric-oxide synthase activity
positive regulation of NF-kappaB transcription factor activity
striated muscle cell differentiation
response to glucocorticoid
response to mineralocorticoid
epithelial tube branching involved in lung morphogenesis
GTPase activity
protein binding
GTP binding
GMP binding
GDP binding
LRR domain binding
protein-containing complex binding
MAPK cascade
positive regulation of protein phosphorylation
stimulatory C-type lectin receptor signaling pathway
epidermal growth factor receptor signaling pathway
Ras protein signal transduction
axon guidance
positive regulation of cell population proliferation
visual learning
positive regulation of gene expression
cytokine-mediated signaling pathway
forebrain astrocyte development
actin cytoskeleton organization
regulation of protein stability
regulation of synaptic transmission, GABAergic
positive regulation of Rac protein signal transduction
social behavior
endocrine signaling
Fc-epsilon receptor signaling pathway
ERBB2 signaling pathway
positive regulation of MAP kinase activity
negative regulation of neuron apoptotic process
negative regulation of cell differentiation
regulation of long-term neuronal synaptic plasticity
homeostasis of number of cells within a tissue
leukocyte migration
positive regulation of nitric-oxide synthase activity
positive regulation of NF-kappaB transcription factor activity
striated muscle cell differentiation
response to glucocorticoid
response to mineralocorticoid
epithelial tube branching involved in lung morphogenesis
GTPase activity
protein binding
GTP binding
GMP binding
GDP binding
LRR domain binding
protein-containing complex binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000256078.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000256078.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000256078.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000256078.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000256078.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000256078.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000256078.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000311936.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000311936.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000311936.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000311936.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000311936.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000311936.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000311936.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000311936.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000311936.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000311936.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000556131.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000556131.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000556131.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000556131.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000556131.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000557334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000557334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000557334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000557334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000557334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000557334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | ENST00000557334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | NM_004985.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | NM_004985.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | NM_004985.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | NM_004985.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | NM_004985.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | NM_004985.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | NM_004985.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | NM_004985.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | NM_004985.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | NM_004985.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | NM_033360.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | NM_033360.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | NM_033360.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | NM_033360.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | NM_033360.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | NM_033360.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | NM_033360.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | NM_033360.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | NM_033360.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | NM_033360.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010158.hg.1 | NM_033360.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000256078.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000256078.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000256078.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000256078.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000256078.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000256078.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000256078.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000311936.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000311936.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000311936.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000311936.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000311936.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000311936.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000311936.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000311936.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000311936.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000311936.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000556131.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000556131.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000556131.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000556131.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000556131.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000557334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000557334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000557334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000557334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000557334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000557334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | ENST00000557334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | NM_004985.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | NM_004985.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | NM_004985.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | NM_004985.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | NM_004985.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | NM_004985.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | NM_004985.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | NM_004985.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | NM_004985.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | NM_004985.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | NM_033360.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | NM_033360.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | NM_033360.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | NM_033360.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | NM_033360.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | NM_033360.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | NM_033360.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | NM_033360.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | NM_033360.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | NM_033360.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010158.hg.1 | NM_033360.3 | Refseq |
