Assay Details
Gene Symbol: TNFAIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006291.2 | NP_006282.2 | 1 | 164 | 92 | |
XM_006720243.3 | XP_006720306.1 | 3 | 805 | 92 | ||
XM_011537112.2 | XP_011535414.1 | 4 | 937 | 92 | ||
XM_011537113.2 | XP_011535415.1 | 3 | 337 | 92 | ||
XM_011537114.2 | XP_011535416.1 | 3 | 806 | 92 | ||
XM_017021616.1 | XP_016877105.1 | 3 | 332 | 92 | ||
GenBank mRNA | BC017818.1 | - | 2 | 223 | 92 | |
BC128449.1 | - | 1 | 102 | 40122621 | 92 | |
BC128450.1 | - | 1 | 102 | 92 | ||
M92357.1 | - | 1 | 164 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC128449.1, M92357.1, NM_006291.2, XM_006720243.3, XM_011537112.2, XM_011537113.2, XM_011537114.2, XM_017021616.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
