Assay Details
Gene Symbol: BAG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015049.2 | NP_001015049.1 | 1 | 241 | 74 | |
GenBank mRNA | BC044216.1 | - | 1 | 200 | 5745133 | 74 |
BI561258.1 | - | 1 | 227 | 74 | ||
DB457402.1 | - | 1 | 222 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC044216.1, NM_001015049.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein folding
Golgi organization
negative regulation of neuron projection development
negative regulation of protein ubiquitination
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
regulation of ubiquitin-protein transferase activity
negative regulation of ubiquitin-protein transferase activity
negative regulation of protein refolding
neuron death
regulation of inclusion body assembly
regulation of cellular response to heat
negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
adenyl-nucleotide exchange factor activity
protein binding
protein kinase binding
ubiquitin protein ligase binding
chaperone binding
protein folding
Golgi organization
negative regulation of neuron projection development
negative regulation of protein ubiquitination
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
regulation of ubiquitin-protein transferase activity
negative regulation of ubiquitin-protein transferase activity
negative regulation of protein refolding
neuron death
regulation of inclusion body assembly
regulation of cellular response to heat
negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
adenyl-nucleotide exchange factor activity
protein binding
protein kinase binding
ubiquitin protein ligase binding
chaperone binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000299204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000299204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000299204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000299204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000299204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000299204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000299204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000337322.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000337322.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000337322.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000337322.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000337322.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000337322.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000337322.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000337322.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000445922.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000445922.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000445922.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000445922.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000445922.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000445922.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000445922.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000557666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000557666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000557666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | ENST00000557666.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_001015048.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_001015048.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_001015048.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_001015048.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_001015048.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_001015048.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_001015048.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_001015049.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_001015049.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_001015049.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_001015049.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_001015049.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_001015049.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_001015049.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_001015049.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_001015049.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_004873.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_004873.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_004873.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_004873.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_004873.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_004873.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010320.hg.1 | NM_004873.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000299204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000299204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000299204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000299204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000299204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000299204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000299204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000337322.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000337322.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000337322.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000337322.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000337322.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000337322.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000337322.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000337322.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000445922.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000445922.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000445922.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000445922.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000445922.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000445922.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000445922.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000557666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000557666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000557666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | ENST00000557666.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_001015048.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_001015048.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_001015048.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_001015048.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_001015048.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_001015048.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_001015048.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_001015049.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_001015049.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_001015049.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_001015049.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_001015049.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_001015049.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_001015049.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_001015049.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_001015049.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_004873.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_004873.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_004873.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_004873.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_004873.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_004873.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010320.hg.1 | NM_004873.3 | Refseq |
