Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000292894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000292894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000292894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000292894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000292894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000292894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000292894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000292894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000292894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000292894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000522483.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000522483.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000522483.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000522483.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000522483.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000522483.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000522483.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000524106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000524106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000524106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000607730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000607730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000607730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000607730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000607730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
ENST00000607730.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
NM_152658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
NM_152658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
NM_152658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
NM_152658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
NM_152658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
NM_152658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
NM_152658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
NM_152658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
NM_152658.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900010510.hg.1 |
NM_152658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000292894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000292894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000292894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000292894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000292894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000292894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000292894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000292894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000292894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000292894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000522483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000522483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000522483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000522483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000522483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000522483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000522483.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000524106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000524106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000524106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000607730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000607730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000607730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000607730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000607730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
ENST00000607730.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
NM_152658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
NM_152658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
NM_152658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
NM_152658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
NM_152658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
NM_152658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
NM_152658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
NM_152658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
NM_152658.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900010510.hg.1 |
NM_152658.2 |
Refseq |