Assay Details
Gene Symbol: DESI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015704.2 | NP_056519.1 | 6 | 2775 | 60 | |
GenBank mRNA | AK090725.1 | - | 2 | 653 | 60 | |
BC008357.1 | - | 1 | 583 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000463886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000463886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000463886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000463886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000463886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000463886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000463886.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000468151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000468151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000468151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000468151.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000482612.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000482612.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | ENST00000482612.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000263256.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000463886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000463886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000463886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000463886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000463886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000463886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000463886.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000468151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000468151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000468151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000468151.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000482612.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000482612.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | ENST00000482612.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | NM_015704.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008832.hg.1 | NM_015704.2 | Refseq |
