Assay Details
Gene Symbol: LINC01096 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_015450.1 | 1 | 354 | 113 | ||
GenBank mRNA | BC035174.2 | - | 1 | 323 | 113 | |
BC048110.1 | - | 1 | 331 | 113 | ||
BC053955.1 | - | 1 | 331 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
