Assay Details
Gene Symbol: LOC100131170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK096606.1 | - | 1 | 318 | 64 | |
AK098403.1 | - | 2 | 1844 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
