Assay Details
Gene Symbol: LOC102725079 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_131941.1 | 3 - 4 | 214 | 96 | ||
GenBank mRNA | AI024749.1 | - | 2 - 3 | 96 | ||
BX094204.1 | - | 3 - 4 | 204 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
