Assay Details

More Information
Additional Information:
This assay is in the 5'UTR region of the AK093312.1, AY517502.1, NM_201594.2, XM_017003834.1, XM_017003835.1, XM_017003836.1, XM_017003837.1, XM_017003838.1, XM_017003839.1, XM_017003840.1, XM_017003841.1, XM_017003842.1, XM_017003846.1, XM_017003847.1, XM_017003848.1, XM_017003849.1, XM_017003851.1, XM_017003853.1, XM_017003854.1, XM_017003855.1, XM_017003856.1, XM_017003857.1, XM_017003858.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
