Assay Details
Gene Symbol: CBX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003655.2 | NP_003646.2 | 5 | 1748 | 56 | |
XM_011525399.2 | XP_011523701.1 | 3 | 3257 | 56 | ||
GenBank mRNA | AF013956.1 | - | 5 | 1745 | 56 | |
AY390430.1 | - | 6 | 775 | 56 | ||
BC014967.1 | - | 6 | 794 | 4856728 | 56 | |
EU439707.1 | - | 5 | 1786 | 56 | ||
KU178422.1 | - | 6 | 775 | 56 | ||
KU178423.1 | - | 5 | 1585 | 56 | ||
U94344.1 | - | 1 | 217 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
